ENA300791221 Enamine

2-chloro-5-(3-methyl-1,2,4-oxadiazol-5-yl)pyridine

  • MDL number MFCD09864545
  • Cas Number : 959240-62-1
  • Molecular Weight : 195.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66526
Availability details for
Quantity
  • Availability
  • Add to Cart