ENA300791224 Enamine

2-carbamoyl-3-nitrobenzoic acid

  • MDL number MFCD08445350
  • Cas Number : 77326-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66529
Availability details for
Quantity
  • Availability
  • Add to Cart