ENA300791230 Enamine

5-(cyclohexylmethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11170352
  • Cas Number : 1042654-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66537

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66537
Availability details for
Quantity
  • Availability
  • Add to Cart