ENA300791239 Enamine

(2-methyl-3-nitrophenyl)hydrazine

  • MDL number MFCD02663158
  • Cas Number : 412925-91-8
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66549
Availability details for
Quantity
  • Availability
  • Add to Cart