ENA300791241 Enamine

3-amino-1-[4-(pyridin-4-ylmethyl)piperazin-1-yl]propan-1-one

  • MDL number MFCD12882849
  • Cas Number : 1179651-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66552

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66552
Availability details for
Quantity
  • Availability
  • Add to Cart