ENA300791254 Enamine

6-amino-1-[4-(3-methylbutanoyl)piperazin-1-yl]hexan-1-one

  • MDL number MFCD16817498
  • Cas Number : 789488-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66569
Availability details for
Quantity
  • Availability
  • Add to Cart