ENA300791257 Enamine

N-(3-hydroxyphenyl)-1-methyl-1H-pyrazole-4-carboxamide

  • MDL number MFCD11168133
  • Cas Number : 1052629-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66572
Availability details for
Quantity
  • Availability
  • Add to Cart