ENA300791264 Enamine

3-[4-(1-aminoethyl)phenyl]-1,3-oxazolidin-2-one

  • MDL number MFCD13196348
  • Cas Number : 1210001-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56857
Availability details for
Quantity
  • Availability
  • Add to Cart