ENA300791266 Enamine

1-[4-(difluoromethoxy)phenyl]-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD08445025
  • Cas Number : 923749-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66587

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66587
Availability details for
Quantity
  • Availability
  • Add to Cart