ENA300791275 Enamine

1-(4-bromophenyl)cyclobutan-1-amine

  • MDL number MFCD09910041
  • Cas Number : 1094218-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-57336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57336
Availability details for
Quantity
  • Availability
  • Add to Cart