ENA300791281 Enamine

2-[5-methyl-2-(thiophen-2-yl)-1,3-oxazol-4-yl]acetonitrile

  • MDL number MFCD09883483
  • Cas Number : 403611-87-0
  • Molecular Weight : 204.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66603
Availability details for
Quantity
  • Availability
  • Add to Cart