ENA300791288 Enamine

2-(5-chloropentyl)-5-phenyl-2H-1,2,3,4-tetrazole

  • MDL number MFCD11645006
  • Cas Number : 1094667-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66612
Availability details for
Quantity
  • Availability
  • Add to Cart