ENA300791292 Enamine

2-amino-N,4-dimethylpentanamide

  • MDL number MFCD09946787
  • Cas Number : 98433-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57921
Availability details for
Quantity
  • Availability
  • Add to Cart