ENA300791293 Enamine

1-(1,4-diazepan-1-yl)-2,2-dimethylpropan-1-one

  • MDL number MFCD09048331
  • Cas Number : 926198-21-2
  • Molecular Weight : 184.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66620
Availability details for
Quantity
  • Availability
  • Add to Cart