ENA300791298 Enamine

2-(2-chlorophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07376371
  • Cas Number : 944275-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192...194
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66633

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66633
Availability details for
Quantity
  • Availability
  • Add to Cart