ENA300791299 Enamine

2-[2-(pyrazin-2-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD00111663
  • Cas Number : 256529-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66635
Availability details for
Quantity
  • Availability
  • Add to Cart