ENA300791300 Enamine

2-amino-2-phenyl-1-(piperidin-1-yl)ethan-1-one

  • MDL number MFCD09941060
  • Cas Number : 24153-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58323
Availability details for
Quantity
  • Availability
  • Add to Cart