ENA300791304 Enamine

3-methylbutane-1-sulfonyl chloride

  • MDL number MFCD09758974
  • Cas Number : 22795-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66646

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66646
Availability details for
Quantity
  • Availability
  • Add to Cart