ENA300791306 Enamine

3,4-dihydro-2H-1,5-benzodioxepin-7-ylmethanamine

  • MDL number MFCD02682040
  • Cas Number : 23475-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66648

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66648
Availability details for
Quantity
  • Availability
  • Add to Cart