ENA300791313 Enamine

1-(4-bromophenyl)-4-methoxybutan-1-one

  • MDL number MFCD12058709
  • Cas Number : 71434-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66668

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66668
Availability details for
Quantity
  • Availability
  • Add to Cart