ENA300791317 Enamine

2-[5-amino-3-(1-benzofuran-2-yl)-1H-pyrazol-1-yl]ethan-1-ol

  • MDL number MFCD11547558
  • Cas Number : 1152926-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66672

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66672
Availability details for
Quantity
  • Availability
  • Add to Cart