ENA300791318 Enamine

3-[(phenylcarbamoyl)amino]butanoic acid

  • MDL number MFCD02113868
  • Cas Number : 6497-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66674
Availability details for
Quantity
  • Availability
  • Add to Cart