ENA300791319 Enamine

5-(methanesulfonamidomethyl)-2-methylfuran-3-carboxylic acid

  • MDL number MFCD12779398
  • Cas Number : 1183819-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66675
Availability details for
Quantity
  • Availability
  • Add to Cart