ENA300791321 Enamine

3-{1-[(4-chlorophenyl)methyl]-1H-indol-3-yl}propanoic acid

  • MDL number MFCD16990629
  • Cas Number : 219544-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66677
Availability details for
Quantity
  • Availability
  • Add to Cart