ENA300791325 Enamine

N-(5-methyl-1,3-thiazol-2-yl)-1,3-thiazolidine-4-carboxamide

  • MDL number MFCD09929739
  • Cas Number : 1104465-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66681
Availability details for
Quantity
  • Availability
  • Add to Cart