ENA300791327 Enamine

N-(2,3-dimethylphenyl)-1,3-thiazolidine-4-carboxamide

  • MDL number MFCD09940252
  • Cas Number : 1101181-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66683
Availability details for
Quantity
  • Availability
  • Add to Cart