ENA300791331 Enamine

1-(4-aminobutyl)-6-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD11911469
  • Cas Number : 1176712-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66688
Availability details for
Quantity
  • Availability
  • Add to Cart