ENA300791332 Enamine

3-(4-aminophenyl)-1-phenylpropan-1-ol

  • MDL number MFCD09035586
  • Cas Number : 196812-87-0
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66689
Availability details for
Quantity
  • Availability
  • Add to Cart