ENA300791335 Enamine

(4-ethoxy-3,5-dimethylphenyl)methanol

  • MDL number MFCD11188447
  • Cas Number : 1038733-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66692
Availability details for
Quantity
  • Availability
  • Add to Cart