ENA300791337 Enamine

3-[(morpholin-4-yl)methyl]benzoic acid hydrochloride

  • MDL number MFCD09064949
  • Cas Number : 137605-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66696
Availability details for
Quantity
  • Availability
  • Add to Cart