ENA300791339 Enamine

2-{[(oxan-2-yl)methyl]sulfanyl}acetic acid

  • MDL number MFCD11647279
  • Cas Number : 933699-73-1
  • Molecular Weight : 190.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66699
Availability details for
Quantity
  • Availability
  • Add to Cart