ENA300791343 Enamine

2-(ethylsulfanyl)-3-methylbutanoic acid

  • MDL number MFCD11136006
  • Cas Number : 1019538-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66707

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66707
Availability details for
Quantity
  • Availability
  • Add to Cart