ENA300791349 Enamine

4-bromo-1-(butan-2-yl)-1H-pyrazole

  • MDL number MFCD12824340
  • Cas Number : 1179304-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66713
Availability details for
Quantity
  • Availability
  • Add to Cart