ENA300791361 Enamine

2-[5-amino-3-(4-bromophenyl)-1H-pyrazol-1-yl]ethan-1-ol

  • MDL number MFCD11645280
  • Cas Number : 1152967-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66732
Availability details for
Quantity
  • Availability
  • Add to Cart