ENA300791367 Enamine

2-(5-bromo-2-methoxyphenyl)pyrrolidine

  • MDL number MFCD02663508
  • Cas Number : 69639-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61687
Availability details for
Quantity
  • Availability
  • Add to Cart