ENA300791371 Enamine

(isothiocyanatomethyl)cyclobutane

  • MDL number MFCD11634967
  • Cas Number : 1095592-84-9
  • Molecular Weight : 127.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66756

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P311,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-66756
pictograms GHS05,GHS06
hstatements H302,H314,H331
Availability details for
Quantity
  • Availability
  • Add to Cart