ENA300791375 Enamine

ethyl 2-[benzyl(2-cyanoethyl)amino]acetate

  • MDL number MFCD16817525
  • Cas Number : 1488-15-9
  • Molecular Weight : 246.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66761
Availability details for
Quantity
  • Availability
  • Add to Cart