ENA300791380 Enamine

3-amino-1-{3-[methyl(phenyl)amino]propyl}thiourea

  • MDL number MFCD10695557
  • Cas Number : 32813-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66766
Availability details for
Quantity
  • Availability
  • Add to Cart