ENA300791383 Enamine

2-(2-methylphenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07376333
  • Cas Number : 115311-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66772
Availability details for
Quantity
  • Availability
  • Add to Cart