ENA300791387 Enamine

5-(2,4-dimethylphenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD10695790
  • Cas Number : 942587-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66777
Availability details for
Quantity
  • Availability
  • Add to Cart