ENA300791388 Enamine

methyl N-(5-amino-2-fluorophenyl)carbamate

  • MDL number MFCD11128932
  • Cas Number : 68621-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66778
Availability details for
Quantity
  • Availability
  • Add to Cart