ENA300791392 Enamine

2-[(2,3-dihydro-1H-inden-5-yl)formamido]acetic acid

  • MDL number MFCD09811298
  • Cas Number : 1016708-59-0
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66783
Availability details for
Quantity
  • Availability
  • Add to Cart