ENA300791393 Enamine

N-(4-aminophenyl)-3-(azepan-1-yl)propanamide

  • MDL number MFCD09052249
  • Cas Number : 937607-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62309
Availability details for
Quantity
  • Availability
  • Add to Cart