ENA300791400 Enamine

4-(2,3-dimethylphenoxy)butanenitrile

  • MDL number MFCD08087461
  • Cas Number : 896657-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66791

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66791
Availability details for
Quantity
  • Availability
  • Add to Cart