ENA300791404 Enamine

6-methyl-3-phenyl-1,3-diazinane-2,4-dione

  • MDL number MFCD02113901
  • Cas Number : 85606-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66797
Availability details for
Quantity
  • Availability
  • Add to Cart