ENA300791409 Enamine

2-(4-methylpiperazin-1-yl)-2-phenylacetic acid

  • MDL number MFCD09718400
  • Cas Number : 347186-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66803
Availability details for
Quantity
  • Availability
  • Add to Cart