ENA300791439 Enamine

5,7-dimethyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD09741805
  • Cas Number : 57414-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66846
Availability details for
Quantity
  • Availability
  • Add to Cart