ENA300791443 Enamine

2-butylcyclopentan-1-one

  • MDL number MFCD00797962
  • Cas Number : 934-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66850
Availability details for
Quantity
  • Availability
  • Add to Cart