ENA300791446 Enamine

(dimethyl-4H-1,2,4-triazol-3-yl)(phenyl)methanamine

  • MDL number MFCD14639062
  • Cas Number : 1250018-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66860
Availability details for
Quantity
  • Availability
  • Add to Cart