ENA300791448 Enamine

1-(4-hydrazinylbenzenesulfonyl)-4-methylpiperazine

  • MDL number MFCD11981326
  • Cas Number : 184708-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66863
Availability details for
Quantity
  • Availability
  • Add to Cart